Learn More
Medchemexpress LLC 2-Chloroacetamide | 79-07-2 | MFCD00008027 | 99.9% | 93.51 | 25 G
SDP
Catalog No. 5000451653
Encompass_Preferred
2-Chloroacetamide is a chemical compound that functions as both a preservative and a herbicide, effective in both upland and paddy fields. It serves as a biocide in various applications including agriculture, glues, paints, and coatings. This compound exerts its action by inhibiting very-long-chain fatty acid elongase and has demonstrated efficacy against paddy field weeds, such as barnyard grass.
  • Acts as a preservative and herbicide.
  • Suitable for upland and paddy fields.
  • Functions as a biocide in agriculture, glues, paints, and coatings.
  • Inhibits very-long-chain fatty acid elongase.
  • Exhibits herbicidal activities against paddy field weeds, such as barnyard grass.
  • Demonstrates antiproliferative activity against certain leukemia cells.
Catalog No. 50-004-51653 Supplier Medchemexpress LLC Supplier No. 25GHYW010629
2-Chloroacetamide is a chemical compound that functions as both a preservative and a herbicide, effective in both upland and paddy fields. It serves as a biocide in various applications including agriculture, glues, paints, and coatings. This compound exerts its action by inhibiting very-long-chain fatty acid elongase and has demonstrated efficacy against paddy field weeds, such as barnyard grass.
  • Acts as a preservative and herbicide.
  • Suitable for upland and paddy fields.
  • Functions as a biocide in agriculture, glues, paints, and coatings.
  • Inhibits very-long-chain fatty acid elongase.
  • Exhibits herbicidal activities against paddy field weeds, such as barnyard grass.
  • Demonstrates antiproliferative activity against certain leukemia cells.