Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroacetamide 98%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
1kg
Packaging:
Glass bottle
Plastic bottle
79-07-2
C2H4ClNO
93.51
MFCD00008027
VXIVSQZSERGHQP-UHFFFAOYSA-N
chloroacetamide,acetamide, 2-chloro,chloracetamide,2-chloroethanamide,microcide,mergal af,chloracetamid,usaf do-29,2-chloracetamide,2-chloro-acetamide
6580
2-chloroacetamide
C(C(=O)N)Cl
79-07-2
C2H4ClNO
93.51
MFCD00008027
VXIVSQZSERGHQP-UHFFFAOYSA-N
chloroacetamide,acetamide, 2-chloro,chloracetamide,2-chloroethanamide,microcide,mergal af,chloracetamid,usaf do-29,2-chloracetamide,2-chloro-acetamide
6580
2-chloroacetamide
C(C(=O)N)Cl
Boiling Point 224.5°C
Packaging Glass bottle
Color White
Density 0.8400g/mL
Quantity 5g
Specific Gravity 0.84
Infrared Spectrum Authentic
Flash Point 170°C
Melting Point 116.0°C to 120.0°C
Water 0.2% max.
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClCH2CONH2
Beilstein 02, 199
Fieser 13,70
Merck Index 15, 2110
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 93.51
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Chloroacetamide, 98%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
Suspected of damaging fertility.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-174-2

RTECSNumber : AB5075000

TSCA : TSCA