Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2'-Chloroacetophenone

Catalog No. 18370925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-370-925G 25 g
18-370-9100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-370-925G Supplier Sigma Aldrich Supplier No. 18370925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2α-chloroacetophenone undergoes stereoselective reduction to (r)-2α-chloro-1-phenyl-ethanol by saccharomyces cerevisiae b5
  • It is commonly used as lacrimator.2α-chloroacetophenone was employed as model substrate to investigate the enzymatic performance of aspergillus terreus and rhizopus oryzae in enantioselective bioreductions using glycerol as a co-solvent

Specifications

CAS 2142-68-9
Density 1.190 g/mL (at 20°C)
Boiling Point 227°C to 230°C
Molecular Formula C8H7ClO
Linear Formula ClC6H4COCH3
MDL Number MFCD00000560
Quantity 25 g
Molecular Weight (g/mol) 154.59
Percent Purity 97%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.