Learn More
Cayman Chemical 2-Chloroadenine, Purity: ≥98%, CAS: 1839-18-5, 250 mg
SDP
Catalog No. 502992140
Encompass_Preferred
A heterocyclic building block; has been used in the synthesis of adenosine antimetabolites with antineoplastic activity; the major catabolite of 2-chloro-2’-deoxyadenosine
Catalog No. 50-299-2140 Supplier Cayman Chemical Supplier No. 30474250 MG
A heterocyclic building block; has been used in the synthesis of adenosine antimetabolites with antineoplastic activity; the major catabolite of 2-chloro-2’-deoxyadenosine