Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroadenine 98.0+%, TCI America™
SDP

Catalog No. C25755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25755G 5 g
C25751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25755G Supplier TCI America Supplier No. C25755G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1839-18-5
Molecular Formula C5H4ClN5
Molecular Weight (g/mol) 169.57
MDL Number MFCD00037925
InChI Key HBJGQJWNMZDFKL-UHFFFAOYSA-N
Synonym 2-chloroadenine, 2-chloro-6-aminopurine, 2-chloro-9h-purin-6-amine, 6-amino-2-chloropurine, adenine, 2-chloro, 6-amino-2-chloro-7h-purine, cladribine impurity c, 1h-purin-6-amine, 2-chloro, 9h-purin-6-amine, 2-chloro, 6-amino-2-chloro-purine
PubChem CID 94904
IUPAC Name 2-chloro-7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC(Cl)=N1

Specifications

Melting Point 389°C
Quantity 5 g
Formula Weight 169.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroadenine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.