Learn More
Cayman Chemical 2-ChloroadenosIn 10mg
SDP
Catalog No. 502969736
Encompass_Preferred
A metabolically stable adenosine analog that binds to adenosine A1, A2A, and A3 receptors with Ki values of 300, 80, and 1,900 nM, respectively; anticonvulsive activity in a rat model of generalized seizures
Catalog No. 50-296-9736 Supplier Cayman Chemical Supplier No. 1561210 MG
A metabolically stable adenosine analog that binds to adenosine A1, A2A, and A3 receptors with Ki values of 300, 80, and 1,900 nM, respectively; anticonvulsive activity in a rat model of generalized seizures