Learn More
Cayman Chemical 2-Chloroadenosine, Purity: ≥98%, CAS: 146-77-0, 50 mg
SDP
Catalog No. 502969739
Encompass_Preferred
A metabolically stable adenosine analog that binds to adenosine A1, A2A, and A3 receptors with Ki values of 300, 80, and 1,900 nM, respectively; anticonvulsive activity in a rat model of generalized seizures
Catalog No. 50-296-9739 Supplier Cayman Chemical Supplier No. 1561250 MG
A metabolically stable adenosine analog that binds to adenosine A1, A2A, and A3 receptors with Ki values of 300, 80, and 1,900 nM, respectively; anticonvulsive activity in a rat model of generalized seizures