Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzaldehyde, 97%

Catalog No. AAA1348830
Change view
Click to view available options
Quantity:
10 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1348830 250 g
AAA1348809 10 g
AAA134880B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1348830 Supplier Thermo Scientific Chemicals Supplier No. A1348830
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 89-98-5 | C7H5ClO | 140.566 g/mol

2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

Solubility
Soluble in water (1.8g/L), alcohol, ether, benzene, and acetone.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-98-5
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.566
MDL Number MFCD00003304
InChI Key FPYUJUBAXZAQNL-UHFFFAOYSA-N
Synonym o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
PubChem CID 6996
IUPAC Name 2-chlorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)Cl

Specifications

Melting Point 10°C to 12°C
Density 1.249
pH 2.9
Boiling Point 212°C
Flash Point 87°C (188°F)
Odor Pungent
Refractive Index 1.5675
Quantity 250 g
UN Number UN3265
Beilstein 385877
Sensitivity Air Sensitive
Solubility Information Soluble in water (1.8g/L),alcohol,ether,benzene,and acetone.
Formula Weight 140.57
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzaldehyde
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 201-956-3
RTECSNumber CU5075000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.