Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzaldehyde, 99%

Catalog No. AC154610050
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154610050 5 mL Glass bottle
AC154611000 100 mL Glass bottle
AC154612500 250 mL Glass bottle
AC154615000 500 mL Glass bottle
AC154610010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC154610050 Supplier Thermo Scientific Chemicals Supplier No. 154610050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 89-98-5 | C7H5ClO | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-98-5
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00003304
InChI Key FPYUJUBAXZAQNL-UHFFFAOYSA-N
Synonym o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
PubChem CID 6996
IUPAC Name 2-chlorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 10°C to 11.5°C
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 209°C to 215°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5650 to 1.567
Linear Formula ClC6H4CHO
Quantity 5 mL
Beilstein 07, 233
Specific Gravity 1.25
Solubility Information Solubility in water: practically insoluble in water
Formula Weight 140.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-956-3
RTECSNumber CU5075000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.