Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzonitrile, 98%

Catalog No. AAB2045130
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2045130 250 g
AAB2045118 50 g
AAB204510B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2045130 Supplier Thermo Scientific Chemicals Supplier No. B2045130
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 873-32-5 | C7H4ClN | 137.566 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-32-5
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.566
MDL Number MFCD00001779
InChI Key NHWQMJMIYICNBP-UHFFFAOYSA-N
Synonym benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech
PubChem CID 13391
IUPAC Name 2-chlorobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)Cl

Specifications

Melting Point 43°C to 46°C
Quantity 250 g
Odor Almond-like
UN Number UN3439
Beilstein 1363916
Boiling Point 232°C
Formula Weight 137.57
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzonitrile
Hazard Category H302+H312-H319
Hazard Statement GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 212-836-5
RTECSNumber DI2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.