Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chlorobenzophenone | 5162-03-8 | MFCD00000558 | 98.8% | 216.66 g/mol | C13H9ClO | 10 G
SDP

Catalog No. 5000304005
Encompass_Preferred

(2-Chlorophenyl)(phenyl)methanone, commonly called 2-chlorobenzophenone, is a chlorinated benzophenone derivative supplied as a high-purity white to off-white solid. It is used in research as a reagent, analytical reference, and synthetic intermediate in organic chemistry.

  • CAS number 5162-03-8.
  • Chemical formula C13H9ClO.
  • Molecular weight 216.66 g/mol.
  • Typical purity 98.8%.
  • Appearance solid, white to off-white.
  • Used as a research reagent, reference standard, and synthetic intermediate.
  • Storage powder: -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-003-04005 Supplier Medchemexpress LLC Supplier No. HYW01411410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Chlorophenyl)(phenyl)methanone, commonly called 2-chlorobenzophenone, is a chlorinated benzophenone derivative supplied as a high-purity white to off-white solid. It is used in research as a reagent, analytical reference, and synthetic intermediate in organic chemistry.

  • CAS number 5162-03-8.
  • Chemical formula C13H9ClO.
  • Molecular weight 216.66 g/mol.
  • Typical purity 98.8%.
  • Appearance solid, white to off-white.
  • Used as a research reagent, reference standard, and synthetic intermediate.
  • Storage powder: -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.