Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzotrichloride 99%, Thermo Scientific™

Catalog No. AC277351000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC277351000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC277351000 Supplier Thermo Scientific Chemicals Supplier No. 277351000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2136-89-2
Molecular Formula C7H4Cl4
Molecular Weight (g/mol) 229.909
InChI Key MFHPYLFZSCSNST-UHFFFAOYSA-N
Synonym 1-chloro-2-trichloromethyl benzene, 2-chlorobenzotrichloride, o-chlorobenzotrichloride, benzene, chloro trichloromethyl, o-chlorobenzylidyne chloride, benzene, 1-chloro-2-trichloromethyl, o-chlorophenyltrichloromethane, alpha,alpha,alpha,2-tetrachlorotoluene, chlorophenyltrichloromethane, alpha,alpha,alpha,ar-tetrachlorotoluene
PubChem CID 16494
IUPAC Name 1-chloro-2-(trichloromethyl)benzene
SMILES C1=CC=C(C(=C1)C(Cl)(Cl)Cl)Cl

Specifications

Color Colorless
CAS Min % 98.5
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Physical Form Liquid After Melting
Refractive Index 1.5820 to 1.5840 (20°C, 589nm)
Linear Formula ClC6H4CCl3
Beilstein 05, 302
Formula Weight 229.92
Percent Purity ≥98.5%
Chemical Name or Material 2-Chlorobenzotrichloride, 99%
Show More Show Less
Hazard Category CANCER SUSPECTED AGENT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.