Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzotrichloride 98.0+%, TCI America™
SDP

Catalog No. C058625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C058625G 25 g
C0586500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C058625G Supplier TCI America Supplier No. C058625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2136-89-2
Molecular Formula C7H4Cl4
Molecular Weight (g/mol) 229.909
MDL Number MFCD00000788
InChI Key MFHPYLFZSCSNST-UHFFFAOYSA-N
Synonym 1-chloro-2-trichloromethyl benzene, 2-chlorobenzotrichloride, o-chlorobenzotrichloride, benzene, chloro trichloromethyl, o-chlorobenzylidyne chloride, benzene, 1-chloro-2-trichloromethyl, o-chlorophenyltrichloromethane, alpha,alpha,alpha,2-tetrachlorotoluene, chlorophenyltrichloromethane, alpha,alpha,alpha,ar-tetrachlorotoluene
PubChem CID 16494
IUPAC Name 1-chloro-2-(trichloromethyl)benzene
SMILES C1=CC=C(C(=C1)C(Cl)(Cl)Cl)Cl

Specifications

Color White
Quantity 25 g
Physical Form Crystal at 20°C
UN Number 1759
Formula Weight 229.91
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chlorobenzotrichloride
EINECSNumber (3)-0089&(3)-0093
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.