Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chlorobenzoxazole | 615-18-9 | MFCD00005766 | 98.5% | 153.57 | C7H4ClNO | 50 G
SDP

Catalog No. 5000299546
Encompass_Preferred

2-Chlorobenzoxazole is an organic biochemical reagent (CAS 615-18-9) commonly used as an intermediate in organic synthesis and as a research reagent in medicinal chemistry and analytical applications. The compound is supplied as a solid with high reported purity and defined storage recommendations for stability.

  • High purity (≈98.5%).
  • Molecular formula C7H4ClNO; molecular weight 153.57.
  • Suitable as an intermediate and research reagent for synthesis and analysis.
  • Available in multiple pack sizes to support small-scale and bulk experiments.
  • Recommended storage at -20°C for long-term stability; follow solvent-specific guidelines for solutions.

Catalog No. 50-002-99546 Supplier Medchemexpress LLC Supplier No. HYW01960650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chlorobenzoxazole is an organic biochemical reagent (CAS 615-18-9) commonly used as an intermediate in organic synthesis and as a research reagent in medicinal chemistry and analytical applications. The compound is supplied as a solid with high reported purity and defined storage recommendations for stability.

  • High purity (≈98.5%).
  • Molecular formula C7H4ClNO; molecular weight 153.57.
  • Suitable as an intermediate and research reagent for synthesis and analysis.
  • Available in multiple pack sizes to support small-scale and bulk experiments.
  • Recommended storage at -20°C for long-term stability; follow solvent-specific guidelines for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.