Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chlorobenzoxazole | 615-18-9 | MFCD00005766 | 98.5% | 153.57 g/mol | C7H4ClNO | 1 ML
SDP

Catalog No. 5000301680
Encompass_Preferred

2-Chlorobenzoxazole is a small-molecule biochemical reagent used in life-science research and organic synthesis. It is supplied as ready-to-use solutions and bulk solids, and comes with analytical and handling documentation to support safe use and identity confirmation.

  • Ready-to-use solution available (10 mM in DMSO, 1 mL).
  • High purity (98.5%).
  • Chemical formula C7H4ClNO; molecular weight 153.57 g/mol.
  • Analytical documents available: SDS, COA, HNMR, GC.
  • Suitable for organic synthesis and biochemical research applications.
  • Stable in DMSO stock solutions; follow handling instructions for concentrated preparations.

Catalog No. 50-003-01680 Supplier Medchemexpress LLC Supplier No. HYW01960610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chlorobenzoxazole is a small-molecule biochemical reagent used in life-science research and organic synthesis. It is supplied as ready-to-use solutions and bulk solids, and comes with analytical and handling documentation to support safe use and identity confirmation.

  • Ready-to-use solution available (10 mM in DMSO, 1 mL).
  • High purity (98.5%).
  • Chemical formula C7H4ClNO; molecular weight 153.57 g/mol.
  • Analytical documents available: SDS, COA, HNMR, GC.
  • Suitable for organic synthesis and biochemical research applications.
  • Stable in DMSO stock solutions; follow handling instructions for concentrated preparations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.