Learn More
Medchemexpress LLC 2-chlorobenzoxazole | 615-18-9 | MFCD00005766 | 98.5% | 153.57 g/mol | C7H4ClNO | 100 G
SDP
Catalog No. 5000300269
Encompass_Preferred
2-Chlorobenzoxazole is an organic biochemical reagent used as a building block and intermediate in medicinal chemistry and organic synthesis. It is a chlorinated benzoxazole derivative with CAS 615-18-9, chemical formula C7H4ClNO, and a molecular weight of 153.57 g/mol. Recommended storage and handling support long-term stability for research use.
  • High purity suitable for research applications (98.5%).
  • Available in solution and bulk solid pack sizes for flexibility.
  • Compatible with common organic solvents for downstream reactions.
  • Stable under recommended cold storage for extended shelf life.
  • Useful as a synthetic intermediate in heterocyclic chemistry.
Catalog No. 50-003-00269 Supplier Medchemexpress LLC Supplier No. HYW019606100G
2-Chlorobenzoxazole is an organic biochemical reagent used as a building block and intermediate in medicinal chemistry and organic synthesis. It is a chlorinated benzoxazole derivative with CAS 615-18-9, chemical formula C7H4ClNO, and a molecular weight of 153.57 g/mol. Recommended storage and handling support long-term stability for research use.
  • High purity suitable for research applications (98.5%).
  • Available in solution and bulk solid pack sizes for flexibility.
  • Compatible with common organic solvents for downstream reactions.
  • Stable under recommended cold storage for extended shelf life.
  • Useful as a synthetic intermediate in heterocyclic chemistry.