Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chlorobenzoxazole | 615-18-9 | MFCD00005766 | 98.5% | 153.57 g/mol | C7H4ClNO | 250 G
SDP

Catalog No. 5000302779
Encompass_Preferred

2-Chlorobenzoxazole (CAS 615-18-9) is a chlorinated benzoxazole used as an organic reagent in chemical synthesis and life-science research. It is provided as a solid or as DMSO solutions for laboratory use and is intended for research purposes only.

  • High purity (≈98.5%).
  • Molecular formula C7H4ClNO; molecular weight 153.57 g/mol.
  • Available as solid quantities and ready-to-use DMSO solutions, up to 250 g.
  • Stable when stored at -20 °C; stock solutions stable at -80 °C for extended storage.
  • Suitable reagent for synthetic chemistry and preparation of biological materials.

Catalog No. 50-003-02779 Supplier Medchemexpress LLC Supplier No. 250GHYW019606
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chlorobenzoxazole (CAS 615-18-9) is a chlorinated benzoxazole used as an organic reagent in chemical synthesis and life-science research. It is provided as a solid or as DMSO solutions for laboratory use and is intended for research purposes only.

  • High purity (≈98.5%).
  • Molecular formula C7H4ClNO; molecular weight 153.57 g/mol.
  • Available as solid quantities and ready-to-use DMSO solutions, up to 250 g.
  • Stable when stored at -20 °C; stock solutions stable at -80 °C for extended storage.
  • Suitable reagent for synthetic chemistry and preparation of biological materials.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.