Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzoxazole, 99%, Thermo Scientific™

Catalog No. AC404530250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404530250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404530250 Supplier Thermo Scientific Chemicals Supplier No. 404530250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-18-9
Molecular Formula C7H4ClNO
Molecular Weight (g/mol) 153.57
MDL Number MFCD00005766
InChI Key BBVQDWDBTWSGHQ-UHFFFAOYSA-N
Synonym 2-chlorobenzoxazole, benzoxazole, 2-chloro, 2-chlorobenzo d oxazole, benzoxazole, chloro, chlorobenzoxazole, 2-chlorobenzoxazol, 2-chlorobenzooxazole, 2-chloro benzoxazole, 2-chloro-benzoxazole, 2-chloro-benzooxazole
PubChem CID 11986
IUPAC Name 2-chloro-1,3-benzoxazole
SMILES ClC1=NC2=CC=CC=C2O1

Specifications

Melting Point 7.0°C
Density 1.3500g/mL
Boiling Point 200.0°C
Flash Point 85°C
Refractive Index 1.5655 to 1.5675
Quantity 25 g
Formula Weight 153.57
Chemical Name or Material 2-Chlorobenzoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.