Learn More
Medchemexpress LLC 2-chlorobiphenyl-2′,3′,4′,5′,6′-d5 | 51624-35-2 | 99.7% | 193.68 | C12H4D5Cl | 1 MG
SDP
Catalog No. 5000302300
Encompass_Preferred
Deuterium-labeled 2-chlorobiphenyl-2′,3′,4′,5′,6′-d5 is an isotopically labeled reference standard used as an internal standard for mass spectrometry and environmental analysis. Supplied as a high-purity material for analytical and research applications; intended for research use only.
  • Deuterium-labeled (d5) isotopologue of 2-chlorobiphenyl.
  • High chemical purity (99.7%) for reliable quantitation.
  • Suitable as an internal standard in GC-MS and LC-MS workflows.
  • Available in small quantitative pack sizes for analytical use.
  • Stable when stored under recommended conditions for long-term use.
Catalog No. 50-003-02300 Supplier Medchemexpress LLC Supplier No. HYW015267S1MG
Deuterium-labeled 2-chlorobiphenyl-2′,3′,4′,5′,6′-d5 is an isotopically labeled reference standard used as an internal standard for mass spectrometry and environmental analysis. Supplied as a high-purity material for analytical and research applications; intended for research use only.
  • Deuterium-labeled (d5) isotopologue of 2-chlorobiphenyl.
  • High chemical purity (99.7%) for reliable quantitation.
  • Suitable as an internal standard in GC-MS and LC-MS workflows.
  • Available in small quantitative pack sizes for analytical use.
  • Stable when stored under recommended conditions for long-term use.