Learn More
Medchemexpress LLC 2-chlorobiphenyl-2′,3′,4′,5′,6′-d5 | 51624-35-2 | 99.7% | 193.68 g·mol⁻¹ | C12H4D5Cl | 5 MG
SDP
Catalog No. 5000303211
Encompass_Preferred
2-Chlorobiphenyl-2′,3′,4′,5′,6′-d5 is the deuterium-labeled isotopologue of 2-chlorobiphenyl (CAS 51624-35-2) supplied for research use as an internal standard and tracer in quantitative analytical methods such as NMR, GC-MS, and LC-MS. It is provided with high chemical purity and defined storage recommendations.
  • Deuterium-labeled (d5) isotopologue.
  • Intended as an internal standard and tracer for NMR, GC-MS, and LC-MS.
  • Chemical purity 99.7% for quantitative analyses.
  • Available in small package sizes suitable for analytical work (1 mg, 5 mg, 10 mg).
  • Recommended storage: -20°C for long-term stability (pure form).
Catalog No. 50-003-03211 Supplier Medchemexpress LLC Supplier No. HYW015267S5MG
2-Chlorobiphenyl-2′,3′,4′,5′,6′-d5 is the deuterium-labeled isotopologue of 2-chlorobiphenyl (CAS 51624-35-2) supplied for research use as an internal standard and tracer in quantitative analytical methods such as NMR, GC-MS, and LC-MS. It is provided with high chemical purity and defined storage recommendations.
  • Deuterium-labeled (d5) isotopologue.
  • Intended as an internal standard and tracer for NMR, GC-MS, and LC-MS.
  • Chemical purity 99.7% for quantitative analyses.
  • Available in small package sizes suitable for analytical work (1 mg, 5 mg, 10 mg).
  • Recommended storage: -20°C for long-term stability (pure form).