Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobiphenyl Solution (BZ No. 1), 100μg/mL in Hexane, Ultra Scientific

Catalog No. USRPC006S
Encompass
Change view
Click to view available options
Quantity:
1 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPC-006S 1 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-RPC-006S Supplier Agilent Technologies Supplier No. RPC006S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2051-60-7 , 110-54-3
Molecular Formula C12H9Cl
Molecular Weight (g/mol) 188.654
InChI Key LAXBNTIAOJWAOP-UHFFFAOYSA-N
PubChem CID 249266
ChEBI CHEBI:34269
IUPAC Name 1-chloro-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2Cl

Specifications

Physical Form Liquid
Color Colorless
Quantity 1 x 2 mL
Concentration or Composition (by Analyte or Components) 100μg/mL (hexane)
Chemical Name or Material 2-Chlorobiphenyl Solution (BZ No. 1), 100μg/mL (hexane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.