Learn More
Medchemexpress LLC 2-chlorocinnamic acid | 3752-25-8 | MFCD00004372 | 99.8% | 182.60 g/mol | C9H7ClO2 | 10 G
SDP
Catalog No. 5000302163
Encompass_Preferred
2-Chlorocinnamic acid (CAS 3752-25-8) is the 2-chloro analogue of cinnamic acid, present predominantly as the trans (E) isomer. It is supplied as a research-grade chemical for use as a building block, synthetic intermediate, and analytical reference in organic and medicinal chemistry; molecular formula C9H7ClO2, molar mass 182.60 g/mol.
  • High purity (≈99.8%).
  • Predominantly trans (E) isomer.
  • Crystalline solid; easy to handle and store.
  • Used as a synthetic building block and intermediate.
  • Suitable as an analytical reference standard and for method development.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-003-02163 Supplier Medchemexpress LLC Supplier No. HYW01636610G
2-Chlorocinnamic acid (CAS 3752-25-8) is the 2-chloro analogue of cinnamic acid, present predominantly as the trans (E) isomer. It is supplied as a research-grade chemical for use as a building block, synthetic intermediate, and analytical reference in organic and medicinal chemistry; molecular formula C9H7ClO2, molar mass 182.60 g/mol.
  • High purity (≈99.8%).
  • Predominantly trans (E) isomer.
  • Crystalline solid; easy to handle and store.
  • Used as a synthetic building block and intermediate.
  • Suitable as an analytical reference standard and for method development.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Documentation available: datasheet, certificate of analysis, and safety data sheet.