Learn More
Medchemexpress LLC 2-chlorocinnamic acid | 3752-25-8 | MFCD00004372 | 99.9% | 182.60 g/mol | C9H7ClO2 | 25 G
SDP
Catalog No. 5000303482
Encompass_Preferred
2-Chlorocinnamic acid is a trans-cinnamate derivative supplied for laboratory research. It is typically encountered as a white to off-white solid with a well-documented CAS number and molecular data; the material is used as a building block or intermediate in small-molecule synthesis and formulation studies.
  • High purity suitable for research applications.
  • White to off-white solid appearance for easy visual identification.
  • Soluble in DMSO (100 mg/mL) for preparation of concentrated stock solutions.
  • Stable under recommended storage conditions with multi-year shelf life as a powder.
  • Documented physicochemical identifiers including CAS, molecular formula, and molecular weight.
Catalog No. 50-003-03482 Supplier Medchemexpress LLC Supplier No. 25GHYW016366
2-Chlorocinnamic acid is a trans-cinnamate derivative supplied for laboratory research. It is typically encountered as a white to off-white solid with a well-documented CAS number and molecular data; the material is used as a building block or intermediate in small-molecule synthesis and formulation studies.
  • High purity suitable for research applications.
  • White to off-white solid appearance for easy visual identification.
  • Soluble in DMSO (100 mg/mL) for preparation of concentrated stock solutions.
  • Stable under recommended storage conditions with multi-year shelf life as a powder.
  • Documented physicochemical identifiers including CAS, molecular formula, and molecular weight.