Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorocyclododecanone 97.0+%, TCI America™
SDP

Catalog No. C16505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16505G 5 g
C16501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16505G Supplier TCI America Supplier No. C16505G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35951-28-1
Molecular Formula C12H21ClO
Molecular Weight (g/mol) 216.749
MDL Number MFCD04117912
InChI Key VDNDILHUSVTMNI-UHFFFAOYSA-N
PubChem CID 566533
IUPAC Name 2-chlorocyclododecan-1-one
SMILES C1CCCCCC(=O)C(CCCC1)Cl

Specifications

Color White
Quantity 5 g
Formula Weight 216.75
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorocyclododecanone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.