Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloroethyl (2-chloroethyl)phosphonate | 17378-30-2 | MFCD24393365 | 206.99 | C4H9Cl2O3P | 50 MG
SDP

Catalog No. 5000285024
Encompass_Preferred

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis and building phosphonate-containing molecules for research applications. It is provided as an analytical-grade reagent; consult the certificate of analysis for storage and characterization details.

  • Organophosphorus synthesis intermediate.
  • Molecular formula C4H9Cl2O3P.
  • Molecular weight 206.99 g/mol.
  • CAS number 17378-30-2.
  • Packaged as a 50 MG unit.
  • Refer to the certificate of analysis for purity and storage instructions.

Catalog No. 50-002-85024 Supplier Medchemexpress LLC Supplier No. HYW58752950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis and building phosphonate-containing molecules for research applications. It is provided as an analytical-grade reagent; consult the certificate of analysis for storage and characterization details.

  • Organophosphorus synthesis intermediate.
  • Molecular formula C4H9Cl2O3P.
  • Molecular weight 206.99 g/mol.
  • CAS number 17378-30-2.
  • Packaged as a 50 MG unit.
  • Refer to the certificate of analysis for purity and storage instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.