Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloroethyl (2-chloroethyl)phosphonate | 17378-30-2 | MFCD24393365 | >95.0% | 206.99 | C4H9Cl2O3P | 10 MG
SDP

Catalog No. 5000285114
Encompass_Preferred

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis. The compound is identified by CAS 17378-30-2 and is characterized by the molecular formula C4H9Cl2O3P and a molecular weight of 206.99 g/mol (manufacturer datasheet).

  • Used as a building block or intermediate in organic synthesis.
  • Organophosphorus chemical class with a phosphoryl functionality.
  • CAS number 17378-30-2 for unambiguous identification.
  • Molecular formula C4H9Cl2O3P, molecular weight 206.99 g/mol.
  • Reported purity >95% (HPLC) from analytical standards vendors.
  • Available in small laboratory quantities suitable for research use.

Catalog No. 50-002-85114 Supplier Medchemexpress LLC Supplier No. HYW58752910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis. The compound is identified by CAS 17378-30-2 and is characterized by the molecular formula C4H9Cl2O3P and a molecular weight of 206.99 g/mol (manufacturer datasheet).

  • Used as a building block or intermediate in organic synthesis.
  • Organophosphorus chemical class with a phosphoryl functionality.
  • CAS number 17378-30-2 for unambiguous identification.
  • Molecular formula C4H9Cl2O3P, molecular weight 206.99 g/mol.
  • Reported purity >95% (HPLC) from analytical standards vendors.
  • Available in small laboratory quantities suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.