Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloroethyl (2-chloroethyl)phosphonate | 17378-30-2 | MFCD24393365 | >95.0% | 206.99 | C4H9Cl2O3P | 5 MG
SDP

Catalog No. 5000283976
Encompass_Preferred

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis. It is a low-molecular-weight phosphonate (C4H9Cl2O3P, MW 206.99) with CAS 17378-30-2, provided with supporting documentation; consult the certificate of analysis for recommended storage and handling.

  • Organophosphorus intermediate for organic synthesis.
  • Molecular formula C4H9Cl2O3P; molecular weight 206.99.
  • CAS number 17378-30-2 for unambiguous identification.
  • Purity greater than 95% by HPLC.
  • Available in small research quantities, such as 5 mg.
  • Supplied with certificate of analysis for storage and handling guidance.

Catalog No. 50-002-83976 Supplier Medchemexpress LLC Supplier No. HYW5875295MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis. It is a low-molecular-weight phosphonate (C4H9Cl2O3P, MW 206.99) with CAS 17378-30-2, provided with supporting documentation; consult the certificate of analysis for recommended storage and handling.

  • Organophosphorus intermediate for organic synthesis.
  • Molecular formula C4H9Cl2O3P; molecular weight 206.99.
  • CAS number 17378-30-2 for unambiguous identification.
  • Purity greater than 95% by HPLC.
  • Available in small research quantities, such as 5 mg.
  • Supplied with certificate of analysis for storage and handling guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.