Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloroethyl (2-chloroethyl)phosphonate | 17378-30-2 | MFCD24393365 | 98.0% | 206.99 | C4H9Cl2O3P | 25 MG
SDP

Catalog No. 5000284861
Encompass_Preferred

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis. It has molecular formula C4H9Cl2O3P and a molecular weight of 206.99 g/mol. Store the material under the conditions recommended in the certificate of analysis and consult the product data sheet for technical details.

  • Organophosphorus compound used as an intermediate in organic synthesis.
  • Molecular formula C4H9Cl2O3P and molecular weight 206.99 g/mol.
  • Contains chloroethyl and phosphonate functional groups.
  • Suitable for small-scale research and synthetic applications.
  • Store under recommended conditions per certificate of analysis.
  • Product data sheet available for technical reference.

Catalog No. 50-002-84861 Supplier Medchemexpress LLC Supplier No. HYW58752925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloroethyl (2-chloroethyl)phosphonate is an organophosphorus compound used as an intermediate in organic synthesis. It has molecular formula C4H9Cl2O3P and a molecular weight of 206.99 g/mol. Store the material under the conditions recommended in the certificate of analysis and consult the product data sheet for technical details.

  • Organophosphorus compound used as an intermediate in organic synthesis.
  • Molecular formula C4H9Cl2O3P and molecular weight 206.99 g/mol.
  • Contains chloroethyl and phosphonate functional groups.
  • Suitable for small-scale research and synthetic applications.
  • Store under recommended conditions per certificate of analysis.
  • Product data sheet available for technical reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.