Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl chloroformate, 97%

Catalog No. AC395331000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395331000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC395331000 Supplier Thermo Scientific Chemicals Supplier No. 395331000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 627-11-2 | C3H4Cl2O2 | 142.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-11-2
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.96
MDL Number MFCD00000646
InChI Key SVDDJQGVOFZBNX-UHFFFAOYSA-N
Synonym 2-chloroethyl chloroformate, chloroethyl chloroformate, carbonochloridic acid, 2-chloroethyl ester, chloroformic acid 2-chloroethyl ester, formic acid, chloro-, 2-chloroethyl ester, chloroethylchloroformate, 2-chloroethylchloroformate, unii-4d32h07z9i, beta-chloroethyl chlorocarbonate, chloroformic acid, beta-chloroethyl ester
PubChem CID 12305
IUPAC Name 2-chloroethyl carbonochloridate
SMILES ClCCOC(Cl)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 1.3900g/mL
Boiling Point 154.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.4470
Linear Formula ClCH2CH2OCOCl
Quantity 100 g
Specific Gravity 1.39
Solubility Information Solubility in water: hydrolyzes
Formula Weight 142.97
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 210-982-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.