Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloroethyl ethyl sulfide

Catalog No. 24264025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-264-025G 25 g
24-264-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-264-025G Supplier Sigma Aldrich Supplier No. 24264025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-chloroethyl ethyl sulfide is a monofunctional analog of sulfur mustard (sm; 2,2α-dichloro diethyl sulfide)
  • The mass diffusivity of 2-chloroethyl ethyl sulfide, a chemical warfare agent simulant, was studied

Specifications

CAS 693-07-2
Density 1.07 g/mL (at 25°C (literature))
Boiling Point 156°C to 157°C (lit.)
Molecular Formula C4H9ClS
Linear Formula ClCH2CH2SC2H5
Refractive Index n20/D 1.4885 (literature)
MDL Number MFCD00000979
Quantity 25 g
Molecular Weight (g/mol) 124.63
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.