Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Laurate 95.0+%, TCI America™
SDP

Catalog No. L010625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L010625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L010625G Supplier TCI America Supplier No. L010625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64919-15-9
Molecular Formula C14H27ClO2
Molecular Weight (g/mol) 262.82
MDL Number MFCD00059475
InChI Key PPRUSMUBWUQYRY-UHFFFAOYSA-N
Synonym Lauric Acid 2-Chloroethyl Ester
PubChem CID 47496
IUPAC Name 2-chloroethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCCl

Specifications

Color Colorless
Boiling Point 187°C
Quantity 25 g
Formula Weight 262.82
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Laurate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.