Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloroethyl phenyl sulfide

Catalog No. 41760225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-760-225ML 25 mL
41-760-25ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-760-225ML Supplier Sigma Aldrich Supplier No. 41760225ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-chloroethyl phenyl sulfide has been reported as sulfur mustard simulant
  • It is a chemical-warfare agent (cwa) simulant and its oxidation using hydrothane films has been reported
  • It is also reported as a simulant for the chemical warfare agent mustard [bis(2-chloroethyl) sulfide]
  • Its reaction with vesicular functionalized double-chain surfactant (potassium o, oα-didodecylphosphorodithioate) at pH 9.0 borate buffer affords [s-[(2-phenylthio)ethyl]o,oα-didodecylphosphorodithioate]
  • Its oxidation with h2o2 catalyzed by [mnl](clo4)2 (L = n,nα,nα-tris(2-hydroxypropyl)-1,4,7-triazacyclononane) afforded 2-chloroethyl phenyl sulfone and phenyl vinyl sulfone.2-chloroethyl phenyl sulfide, a sulfur mustard agent stimulant, may be used as target analyte in a bioassay for the detection of mustard agent

Specifications

CAS 5535-49-9
Density 1.174 g/mL (at 25°C (literature))
Boiling Point 90°C to 91°C (1 mmHg)
Molecular Formula C8H9ClS
Refractive Index n20/D 1.583 (literature)
Linear Formula ClCH2CH2SC6H5
MDL Number MFCD00013691
Quantity 25 mL
Synonym 2-(Phenylthio)ethyl chloride
Molecular Weight (g/mol) 172.68
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.