Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chloroethyl vinyl ether

Catalog No. 10998325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-998-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-998-325G Supplier Sigma Aldrich Supplier No. 10998325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chloroethyl vinyl ether

Specifications

CAS 110-75-8
Density 1.048 g/mL (at 25°C)
Boiling Point 109°C
Molecular Formula C4H7ClO
Refractive Index n20/D 1.438 (literature)
Linear Formula ClCH2CH2OCH=CH2
Quantity 25 g
MDL Number MFCD00000973
Synonym (2-Chloroethoxy)ethene; 1-Chloro-2-vinyloxyethane; 2-Chloroethyl vinyl ether; 2-Vinyloxyethyl chloride; Vinyl 2-chloroethyl ether
Molecular Weight (g/mol) 106.55
Percent Purity 99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.