Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Vinyl Ether (For EPA 600, 8000/Appendix IX Methods)

Catalog No. USHC0701
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HC-070-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HC-070-1 Supplier Agilent Technologies Supplier No. HC0701
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Ampule
CAS 110-75-8
Methods For EPA 600, 8000/ Appendix IX Methods
Molecular Formula C4H7ClO
InChI Key DNJRKFKAFWSXSE-UHFFFAOYSA-N
SMILES C=COCCCl
IUPAC Name 1-chloro-2-ethenoxyethane
Molecular Weight (g/mol) 106.549
Quantity 1 x 1 mL
PubChem CID 8074
Chemical Name or Material 2-Chloroethyl Vinyl Ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.