Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethylphosphoryl Dichloride, TCI America™
SDP

Catalog No. C017125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C017125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C017125G Supplier TCI America Supplier No. C017125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1455-05-6
Molecular Formula C2H4Cl3O2P
Molecular Weight (g/mol) 197.376
MDL Number MFCD00013898
InChI Key YRTAUZXAHQPFJV-UHFFFAOYSA-N
Synonym 2-Chloroethylphosphoric Acid Dichloride, 2-Chloroethyl Dichlorophosphate
PubChem CID 548448
IUPAC Name 1-chloro-2-dichlorophosphoryloxyethane
SMILES C(CCl)OP(=O)(Cl)Cl

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
UN Number 3265
Formula Weight 197.38
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloroethylphosphoryl Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.