Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorohydroquinone, tech. 90%, Thermo Scientific™

Catalog No. AAA1955436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1955436 500 g
AAA1955414 25 g
AAA1955422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1955436 Supplier Thermo Scientific Chemicals Supplier No. A1955436

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-67-8
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.554
MDL Number MFCD00002341
InChI Key AJPXTSMULZANCB-UHFFFAOYSA-N
Synonym chlorohydroquinone, 2-chlorohydroquinone, monochlorohydroquinone, chloroquinol, 1,4-benzenediol, 2-chloro, 2-chloro-1,4-benzenediol, hydroquinone, chloro, 2-chloro-1,4-dihydroxybenzene, 2,5-dihydroxychlorobenzene, 1,4-dihydroxy-2-chlorobenzene
PubChem CID 301
ChEBI CHEBI:27675
IUPAC Name 2-chlorobenzene-1,4-diol
SMILES C1=CC(=C(C=C1O)Cl)O

Specifications

Melting Point 97°C to 102°C
Boiling Point 262°C to 263°C
Quantity 500 g
Formula Weight 144.56
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Chlorohydroquinone
Hazard Category H311-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-442-8
RTECSNumber MX4800000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.