Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Chloroisonicotinic acid | 6313-54-8 | 98.8% | C6H4ClNO2 | 50 G
SDP

Catalog No. 5000434406
Encompass_Preferred

2-Chloroisonicotinic acid is a biochemical reagent suitable for life science related research as a biological material or organic compound. It is intended for research use only and not for sale to patients. This solid compound is off-white to light yellow in appearance and has a molecular weight of 157.55. It is stable under recommended storage conditions.

  • Used as a biochemical reagent
  • Suitable for life science related research
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-34406 Supplier Medchemexpress LLC Supplier No. HY6500350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloroisonicotinic acid is a biochemical reagent suitable for life science related research as a biological material or organic compound. It is intended for research use only and not for sale to patients. This solid compound is off-white to light yellow in appearance and has a molecular weight of 157.55. It is stable under recommended storage conditions.

  • Used as a biochemical reagent
  • Suitable for life science related research
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 100 mg/mL
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.