Learn More
Medchemexpress LLC 2-(Chloromethyl)pyridine hydrochloride | 6959-47-3 | 164.03 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00750210G
Encompass_Preferred
2-(Chloromethyl)pyridine hydrochloride is an alkylating agent used in organic synthesis reactions. It can introduce pyridine groups in reactions with calixarenes, with the product's conformation affected by the base used. It also functions as an electrophile in the alkylation reaction of p-tert-butylcalixarene, resulting in the generation of various pyridinyl homologues.
  • Alkylating agent
  • Used in organic synthesis reactions
  • Introduces pyridine groups in reactions with calixarenes
  • Product conformation influenced by the base used
  • Electrophile in the alkylation reaction of p-tert-butylcalixarene
  • Generates various pyridinyl homologues
  • For research use only
Catalog No. 50-004-46339