Learn More
Medchemexpress LLC 2-(Chloromethyl)pyridine hydrochloride | 6959-47-3 | 164.03 | 100 G
SDP
Catalog No. 5000446338
Encompass_Preferred
2-(Chloromethyl)pyridine hydrochloride is an alkylating agent primarily utilized in organic synthesis reactions. It is intended for research use only and not for patient administration.
  • Used as an alkylating agent in organic synthesis
  • Introduces pyridine groups in reactions with calixarenes
  • Functions as an electrophile in p-tert-butylcalixarene alkylation
Catalog No. 50-004-46338 Supplier Medchemexpress LLC Supplier No. HYW007502100G
2-(Chloromethyl)pyridine hydrochloride is an alkylating agent primarily utilized in organic synthesis reactions. It is intended for research use only and not for patient administration.
  • Used as an alkylating agent in organic synthesis
  • Introduces pyridine groups in reactions with calixarenes
  • Functions as an electrophile in p-tert-butylcalixarene alkylation