Learn More
Medchemexpress LLC 2-(Chloromethyl)pyridine hydrochloride | 6959-47-3 | 164.03 | 25 G
SDP
Catalog No. 5000449852
Encompass_Preferred
2-(Chloromethyl)pyridine hydrochloride is an alkylating agent used in organic synthesis reactions. It can introduce pyridine groups in reactions with calixarenes, with the product's conformation influenced by the base used. This compound also functions as an electrophile in the alkylation of p-tert-butylcalixarene, facilitating the generation of diverse pyridinyl homologues.
  • Functions as an alkylating agent
  • Suitable for organic synthesis reactions
  • Introduces pyridine groups in calixarene reactions
  • Product conformation affected by base used
  • Acts as an electrophile in p-tert-butylcalixarene alkylation
  • Capable of generating various pyridinyl homologues
Catalog No. 50-004-49852 Supplier Medchemexpress LLC Supplier No. 25GHYW007502
2-(Chloromethyl)pyridine hydrochloride is an alkylating agent used in organic synthesis reactions. It can introduce pyridine groups in reactions with calixarenes, with the product's conformation influenced by the base used. This compound also functions as an electrophile in the alkylation of p-tert-butylcalixarene, facilitating the generation of diverse pyridinyl homologues.
  • Functions as an alkylating agent
  • Suitable for organic synthesis reactions
  • Introduces pyridine groups in calixarene reactions
  • Product conformation affected by base used
  • Acts as an electrophile in p-tert-butylcalixarene alkylation
  • Capable of generating various pyridinyl homologues