Learn More
Medchemexpress LLC 2-(Chloromethyl)pyridine hydrochloride | 6959-47-3 | 164.03 | 250 G
SDP
Catalog No. 5000449879
Encompass_Preferred
2-(Chloromethyl)pyridine hydrochloride is an alkylating agent primarily used in organic synthesis reactions. It is known for its ability to introduce pyridine groups in reactions with calixarenes, where the resulting conformation is influenced by the base used. This compound also functions as an electrophile in the alkylation of p-tert-butylcalixarene, facilitating the generation of various pyridinyl homologues. It appears as a yellow to brown solid with a high purity of 99.88% and is intended for research use only.
  • Alkylating agent for organic synthesis
  • Introduces pyridine groups in calixarene reactions
  • Electrophile for alkylation of p-tert-butylcalixarene
  • Generates various pyridinyl homologues
  • Solid, yellow to brown appearance
  • High purity of 99.88%
  • Intended for research use only
Catalog No. 50-004-49879 Supplier Medchemexpress LLC Supplier No. 250GHYW007502
2-(Chloromethyl)pyridine hydrochloride is an alkylating agent primarily used in organic synthesis reactions. It is known for its ability to introduce pyridine groups in reactions with calixarenes, where the resulting conformation is influenced by the base used. This compound also functions as an electrophile in the alkylation of p-tert-butylcalixarene, facilitating the generation of various pyridinyl homologues. It appears as a yellow to brown solid with a high purity of 99.88% and is intended for research use only.
  • Alkylating agent for organic synthesis
  • Introduces pyridine groups in calixarene reactions
  • Electrophile for alkylation of p-tert-butylcalixarene
  • Generates various pyridinyl homologues
  • Solid, yellow to brown appearance
  • High purity of 99.88%
  • Intended for research use only