Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(Chloromethyl)pyrimidine hydrochloride | 936643-80-0 | 99.4% | C5H6Cl2N2 | 50 MG
SDP

Catalog No. 5000357789
Encompass_Preferred

2-(Chloromethyl)pyrimidine hydrochloride is a synthetic intermediate useful for pharmaceutical synthesis. It is sold for research purposes only and not for patient use.

  • Molecular weight: 165.02
  • Appearance: Solid
  • Color: Light brown to yellow
  • Purity: 99.40%

Catalog No. 50-003-57789 Supplier Medchemexpress LLC Supplier No. HY5210250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(Chloromethyl)pyrimidine hydrochloride is a synthetic intermediate useful for pharmaceutical synthesis. It is sold for research purposes only and not for patient use.

  • Molecular weight: 165.02
  • Appearance: Solid
  • Color: Light brown to yellow
  • Purity: 99.40%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.