Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloromethylbenzimidazole, 95%

Catalog No. AC109390050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109390050 5 g
AC109390250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109390050 Supplier Thermo Scientific Chemicals Supplier No. 109390050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4857-04-9 | C8H7ClN2 | 166.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4857-04-9
Molecular Formula C8H7ClN2
Molecular Weight (g/mol) 166.61
MDL Number MFCD00005602
InChI Key SPMLMLQATWNZEE-UHFFFAOYSA-N
IUPAC Name 2-(chloromethyl)-1H-1,3-benzodiazole
SMILES ClCC1=NC2=CC=CC=C2N1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 145°C to 150°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 166.61
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Chloromethylbenzimidazole, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
EINECSNumber 225-452-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.