Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-chloronaphthalene | 91-58-7 | MFCD00035731 | 99.7% | 162.62 | C10H7Cl | 5G
SDP

Supplier:  Medchemexpress LLC HY1244515G

Encompass_Preferred

2-Chloronaphthalene (CAS 91-58-7) is a chlorinated aromatic hydrocarbon supplied as a solid, white to off-white crystalline material. It is used as a reagent and reference standard in organic synthesis and analytical chemistry. The material is offered in a 5 g pack and has high stated purity suitable for laboratory use.

  • High purity of 99.7%.
  • Molecular formula C10H7Cl.
  • Molecular weight 162.62 g·mol-1.
  • Appearance solid, white to off-white.
  • Pack size 5 g.
  • Typical applications include organic synthesis and analytical reference standards.

Catalog No. 50-002-38910


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.