Learn More
Medchemexpress LLC 2-chloronaphthalene-d7 | 93951-84-9 | MFCD00209698 | 98.0% | 169.66 g/mol | C10D7Cl | 1MG
SDP
Catalog No. 5000238909
Encompass_Preferred
2-Chloronaphthalene-d7 is the deuterium-labeled analogue of 2-chloronaphthalene for research use as an isotopically labeled tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS. The compound is supplied in small milligram quantities and is intended for analytical and method development workflows; isotopic enrichment is reported at about 98 atom % D.
  • Deuterium labeling (d7) for isotopic tracing and quantitation
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS
  • Available in small milligram package sizes for analytical workflows
  • High isotopic enrichment (~98 atom % D) for reliable mass discrimination
  • Stable when stored as a powder at low temperatures under recommended conditions
Catalog No. 50-002-38909 Supplier Medchemexpress LLC Supplier No. HY124451S1MG
2-Chloronaphthalene-d7 is the deuterium-labeled analogue of 2-chloronaphthalene for research use as an isotopically labeled tracer and internal standard in quantitative analyses by NMR, GC-MS, and LC-MS. The compound is supplied in small milligram quantities and is intended for analytical and method development workflows; isotopic enrichment is reported at about 98 atom % D.
  • Deuterium labeling (d7) for isotopic tracing and quantitation
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS
  • Available in small milligram package sizes for analytical workflows
  • High isotopic enrichment (~98 atom % D) for reliable mass discrimination
  • Stable when stored as a powder at low temperatures under recommended conditions