Learn More
Medchemexpress LLC 2-Chloronicotinaldehyde | 36404-88-3 | 99.76% | 141.56 | 100 G
SDP
Catalog No. 5000403536
Encompass_Preferred
2-Chloropyridine-3-carbaldehyde, also known as 2-Chloronicotinaldehyde, is a biochemical reagent. It is suitable for use as a biological material or organic compound in life science related research applications.
  • Can be used as a biological material
  • Can be used as an organic compound for life science research
  • Has cytotoxicity against mouse B16F10 cells with an IC50 of 70.74 μM
Catalog No. 50-004-03536 Supplier Medchemexpress LLC Supplier No. HY32791100G
2-Chloropyridine-3-carbaldehyde, also known as 2-Chloronicotinaldehyde, is a biochemical reagent. It is suitable for use as a biological material or organic compound in life science related research applications.
  • Can be used as a biological material
  • Can be used as an organic compound for life science research
  • Has cytotoxicity against mouse B16F10 cells with an IC50 of 70.74 μM