Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloronicotinamide, 98+%, Thermo Scientific™

Catalog No. AAA1209006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1209006 5 g
AAA1209003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1209006 Supplier Thermo Scientific Chemicals Supplier No. A1209006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10366-35-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00006237
InChI Key ZQZAHPFFZWEUCL-UHFFFAOYSA-N
Synonym 2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
PubChem CID 82588
IUPAC Name 2-chloropyridine-3-carboxamide
SMILES C1=CC(=C(N=C1)Cl)C(=O)N

Specifications

Melting Point 163°C to 167°C
Quantity 5 g
Beilstein 119026
Formula Weight 156.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloronicotinamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-808-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.