Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloronicotinoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. C16045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16045G 5 g
C160425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16045G Supplier TCI America Supplier No. C16045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 49609-84-9
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 176.00
MDL Number MFCD00051677
InChI Key RXTRRIFWCJEMEL-UHFFFAOYSA-N
Synonym 2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride
PubChem CID 2774541
IUPAC Name 2-chloropyridine-3-carbonyl chloride
SMILES ClC(=O)C1=CC=CN=C1Cl

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 120°C
Quantity 5 g
UN Number 3261
Formula Weight 176.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloronicotinoyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.