Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenethylalcohol, 98%

Catalog No. AC186190050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186190050 5 g Glass bottle
AC186190010 1 g Glass bottle
AC186190250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC186190050 Supplier Thermo Scientific Chemicals Supplier No. 186190050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 19819-95-5 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19819-95-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00002888
InChI Key IWNHTCBFRSCBQK-UHFFFAOYSA-N
Synonym 2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol
PubChem CID 88266
IUPAC Name 2-(2-chlorophenyl)ethanol
SMILES C1=CC=C(C(=C1)CCO)Cl

Specifications

Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Density 1.1900g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5495 to 1.5515
Specific Gravity 1.19
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4CH2CH2OH
Boiling Point 84.0°C to 85.0°C (3.0 mmHg)
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 2-Chlorophenethylalcohol, 98%
Show More Show Less
EINECSNumber 243-348-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.