Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Chlorophenyl phosphorodichloridate

Catalog No. 23523725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-523-725G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-523-725G Supplier Sigma Aldrich Supplier No. 23523725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Chlorophenyl phosphorodichloridate

Specifications

CAS 15074-54-1
Density 1.491 g/mL (at 25°C (literature))
Boiling Point 135°C to 137°C (12 mmHg)
Molecular Formula C6H4Cl3O2P
Linear Formula Cl2P(O)OC6H4Cl
Refractive Index n20/D 1.538 (literature)
MDL Number MFCD00009704
Quantity 500 mg
Synonym 2-Chlorophenyl dichlorophosphate
Molecular Weight (g/mol) 245.43
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.